Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SMA-560 Mouse Orthotopic Glioma Cell Line

Catalog No. SCC179MI Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Cells:
≥ 1 x 106 Cells/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Cells
SCC179MI ≥ 1 x 106 Cells/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC179MI Supplier MilliporeSigma™ Supplier No. SCC179
Add to Cart
Add to Cart

SMA-560 cell line is a well-established glioma model derived from spontaneous murine astrocytoma

  • Uses:
    • Cell culture
  • Quality Assurance Tests:
    • Cells are tested negative for infectious diseases by a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services
    • Cells are verified to be of mouse origin and negative for inter-species contamination from rat, Chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by Contamination Clear panel by Charles River Animal Diagnostic Services
    • Cells are negative for mycoplasma contamination
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen, Can be cultured for at least 10 passages after initial thawing without affecting the cell marker expression and functionality
Cell Type Cancer Cells
For Use With (Application) Cell Culture
No. of Cells ≥ 1 x 106 Cells/Vial

For research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.