Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human SMAC Protein, His Tag, >95% Purity, 1 mg

Catalog No. 501658874
Encompass_Preferred

Human SMAC, also known as DIABLO, is a mitochondrial protein that acts as an activator of caspase by opposing the inhibitory activity of inhibitor of apoptosis proteins (IAP). This protein promotes apoptosis by activating caspases in the cytochrome c/Apaf-1/caspase-9 pathway and inhibits BIRC6/bruce activity by preventing its binding to caspases. This recombinant protein is expressed in E. coli cells and contains amino acids Ala 56 - Asp 239 with a C-terminal polyhistidine tag.

  • Greater than 95% purity as determined by SDS-PAGE.
  • Calculated molecular weight of 21.7 kDa.
  • Lyophilized from a 0.22 μm filtered solution in 20 mM HEPES, 150 mM NaCl, pH 7.5 with trehalose.
  • Stable for 12 months in lyophilized state at -20°C to -70°C.
  • Validated for technical research applications.

Catalog No. 50-165-8874 Supplier ACROBiosystems Supplier No. DIOH51291MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Human SMAC, also known as DIABLO, is a mitochondrial protein that acts as an activator of caspase by opposing the inhibitory activity of inhibitor of apoptosis proteins (IAP). This protein promotes apoptosis by activating caspases in the cytochrome c/Apaf-1/caspase-9 pathway and inhibits BIRC6/bruce activity by preventing its binding to caspases. This recombinant protein is expressed in E. coli cells and contains amino acids Ala 56 - Asp 239 with a C-terminal polyhistidine tag.

  • Greater than 95% purity as determined by SDS-PAGE.
  • Calculated molecular weight of 21.7 kDa.
  • Lyophilized from a 0.22 μm filtered solution in 20 mM HEPES, 150 mM NaCl, pH 7.5 with trehalose.
  • Stable for 12 months in lyophilized state at -20°C to -70°C.
  • Validated for technical research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.