Learn More
Signosis Inc SMAD/TGFbeta Luciferase Reporter NIH/3T3 Stable Cell Line SL-0030
Catalog No. NC2942624
Encompass
SMAD 2/3 Responsive Luciferase Reporter NIH/3T3 Stable Cell Line is derived from mouse fibroblast,and stably express firefly luciferase reporter gene under the control of SMAD 2/3 response element. This cell line is an ideal cellular model for monitoring the activation of TGFbeta Signaling Pathway triggered by stimuli treatment, enforced gene expression and gene knockdown.
Catalog No. NC2942624 Supplier Signosis Inc Supplier No. SL0030
SMAD 2/3 Responsive Luciferase Reporter NIH/3T3 Stable Cell Line is derived from mouse fibroblast,and stably express firefly luciferase reporter gene under the control of SMAD 2/3 response element. This cell line is an ideal cellular model for monitoring the activation of TGFbeta Signaling Pathway triggered by stimuli treatment, enforced gene expression and gene knockdown.