Learn More
Aviva Systems Biology Corp SMAD1 Antibody - N-terminal region (ARP38692_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38692_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SMAD1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SMAD1. Alternate names: BSP1, JV4-1, JV41, MADH1, MADR1, BSP-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56757