Learn More
Aviva Systems Biology Corp SMAD1 Antibody - middle region (ARP32003_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32003_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMAD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SMAD1. Alternate names: BSP1, JV4-1, JV41, MADH1, MADR1, BSP-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54416