Learn More
Abcam SMAD1 (pS463/465) ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1860361X96TESTS
Encompass_Preferred
SMAD1 (pS463/465) ELISA Kit is a single-wash 90-min Simplestep used to quantify SMAD1 (pS463/S465) with a sensitivity of 10 µg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Semi-quantitative sandwich ELISA - 450 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1791178