Learn More
Aviva Systems Biology Corp SMAD2 Antibody - N-terminal region (ARP38694_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38694_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SMAD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMAD2. Alternate names: JV18, MADH2, MADR2, JV18-1, hMAD-2, hSMAD2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56759