Learn More
Aviva Systems Biology Corp SMAD2 Antibody - C-terminal region (ARP38695_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38695_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMAD2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human SMAD2. Alternate names: JV18, JV18-1, MADH2, MADR2, MGC22139, MGC34440, hMAD-2, hSMAD2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-753