Learn More
Medchemexpress LLC SMAD2 Human His-SU 10ug
SDP
Catalog No. 5000196764
Encompass_Preferred
Recombinant human SMAD2 (His-SUMO) is a purified, E. coli-expressed fragment of human SMAD2 (residues S2-S467) provided as a 10 μg aliquot for research applications such as biochemical characterization, binding studies, and assay development.
  • Purified protein representing residues S2-S467.
  • N-terminal SUMO and 6xHis tag for affinity purification and detection.
  • High purity suitable for biochemical and binding assays.
  • Supplied in stabilizing buffer: 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
  • Small aliquot format (10 μg) for assay optimization and pilot studies.
  • Molecular weight ~68.2 kDa as observed by SDS-PAGE.
Catalog No. 50-001-96764 Supplier Medchemexpress LLC Supplier No. HYP7153610UG
Recombinant human SMAD2 (His-SUMO) is a purified, E. coli-expressed fragment of human SMAD2 (residues S2-S467) provided as a 10 μg aliquot for research applications such as biochemical characterization, binding studies, and assay development.
  • Purified protein representing residues S2-S467.
  • N-terminal SUMO and 6xHis tag for affinity purification and detection.
  • High purity suitable for biochemical and binding assays.
  • Supplied in stabilizing buffer: 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0.
  • Small aliquot format (10 μg) for assay optimization and pilot studies.
  • Molecular weight ~68.2 kDa as observed by SDS-PAGE.