Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SMAD2 Human His-SU 50ug
SDP

Catalog No. 5000195919
Encompass_Preferred

Recombinant human SMAD2 protein (residues S2-S467) expressed in E. coli and supplied as an N-SUMO-6xHis tagged fusion. Supplied lyophilized from 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0, it has an approximate molecular weight of 68.2 kDa and is ≥95% pure, making it suitable for biochemical assays, structural studies, and antibody validation.

  • Recombinant human sequence S2-S467.
  • N-terminal SUMO and 6xHis affinity tag.
  • Expressed in E. coli for efficient production.
  • Lyophilized formulation with trehalose for stability.
  • High purity confirmed by reducing SDS-PAGE.
  • Reconstitution at ≥100 μg/mL recommended for use.

Catalog No. 50-001-95919 Supplier Medchemexpress LLC Supplier No. HYP7153650UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human SMAD2 protein (residues S2-S467) expressed in E. coli and supplied as an N-SUMO-6xHis tagged fusion. Supplied lyophilized from 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose, pH 8.0, it has an approximate molecular weight of 68.2 kDa and is ≥95% pure, making it suitable for biochemical assays, structural studies, and antibody validation.

  • Recombinant human sequence S2-S467.
  • N-terminal SUMO and 6xHis affinity tag.
  • Expressed in E. coli for efficient production.
  • Lyophilized formulation with trehalose for stability.
  • High purity confirmed by reducing SDS-PAGE.
  • Reconstitution at ≥100 μg/mL recommended for use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.