Learn More
Medchemexpress LLC SMAD2 Human His-SU 5ug
SDP
Catalog No. 5000196572
Encompass_Preferred
Recombinant human SMAD2 protein (residues S2-S467) expressed in E. coli and supplied as a lyophilized powder with N-SUMO and N-6xHis tags. The product is accompanied by quality control data including SDS-PAGE purity and endotoxin testing, and includes recommended reconstitution and storage instructions for research use.
  • Lyophilized recombinant protein (S2-S467) with N-SUMO and N-6xHis tags.
  • Expressed in E. coli for high yield and consistent quality.
  • Purity >95% by reducing SDS-PAGE (batch example 97%).
  • Low endotoxin (<1 EU/μg) suitable for sensitive assays.
  • Formulated with trehalose and EDTA for stability as a lyophilized powder.
  • Recommended reconstitution at ≥100 μg/mL; aliquot and freeze for long-term storage.
Catalog No. 50-001-96572 Supplier Medchemexpress LLC Supplier No. HYP715365UG
Recombinant human SMAD2 protein (residues S2-S467) expressed in E. coli and supplied as a lyophilized powder with N-SUMO and N-6xHis tags. The product is accompanied by quality control data including SDS-PAGE purity and endotoxin testing, and includes recommended reconstitution and storage instructions for research use.
  • Lyophilized recombinant protein (S2-S467) with N-SUMO and N-6xHis tags.
  • Expressed in E. coli for high yield and consistent quality.
  • Purity >95% by reducing SDS-PAGE (batch example 97%).
  • Low endotoxin (<1 EU/μg) suitable for sensitive assays.
  • Formulated with trehalose and EDTA for stability as a lyophilized powder.
  • Recommended reconstitution at ≥100 μg/mL; aliquot and freeze for long-term storage.