Learn More
Aviva Systems Biology Corp SMAD4 Antibody - middle region (ARP32005_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32005_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SMAD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SMAD4. Alternate names: JIP, DPC4, MADH4, MYHRSApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54418