Learn More
Aviva Systems Biology Corp SMAD4 Antibody - N-terminal region (ARP38605_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38605_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMAD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMAD4. Alternate names: DPC4, JIP, MADH4, MYHRSApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-56724