Learn More
Aviva Systems Biology Corp SMAD5 Antibody - middle region (ARP32006_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32006_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SMAD5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SMAD5. Alternate names: DWFC, JV5-1, MADH5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54420