Learn More
Aviva Systems Biology Corp SMAD7 Antibody - N-terminal region (ARP32008_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32008_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SMAD7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMAD7. Alternate names: FLJ16482, MADH7, MADH8, CRCS3ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54422