Learn More
Proteintech Group Inc SMAD7 Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 258401AP150UL
Encompass_Preferred
The SMAD7 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SMAD7. This antibody recognizes human, mouse, rat antigen. The SMAD7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Catalog No. 50-225-6448