Learn More
New England Biolabs, Inc. SmaI – 10000 units
SDP
Catalog No. 50812010
Encompass
A restriction endonuclease that recognizes the sequence CCC_^GGG.
Catalog No. 50-812-010 Supplier New England Biolabs, Inc. Supplier No. R0141L
A restriction endonuclease that recognizes the sequence CCC_^GGG.