Learn More
New England Biolabs, Inc. SmaI – 2000 units
SDP
Catalog No. 50812011
Encompass
A restriction endonuclease that recognizes the sequence CCC_^GGG.
Catalog No. 50-812-011 Supplier New England Biolabs, Inc. Supplier No. R0141S
A restriction endonuclease that recognizes the sequence CCC_^GGG.