Learn More
Chemglass Life Sciences Clamp, Three-Prong Extension, Small, Single Adjust, 6.3in OAL
SDP
Catalog No. CG920501
Encompass_Preferred
Clamp, Three-Prong Extension, Small, Single Adjust, 6 1/2in OAL. Nickel-plated zinc alloy clamp has high tensile strength and corrosion resistance. Fully enclosed stainless steel springs provide smooth operation. Three-prong clamps are available in single and dual adjustment. The small clamp has a 1¿ (25mm) maximum grip size with a 1/4¿ (6mm) O.D. extension arm. The medium clamp has a 1 1/2¿ (38mm) maximum grip size with a 1/2¿ (13mm) O.D. extension arm. The large clamp has a 2 1/2¿ (64mm) maximum grip size with a 5/8¿ (16mm) O.D. extension arm. Complete with removable vinyl and fiberglass covers for easy cleaning.
Catalog No. CG920501 Supplier Chemglass Life Sciences Supplier No. CG920501
Clamp, Three-Prong Extension, Small, Single Adjust, 6 1/2in OAL. Nickel-plated zinc alloy clamp has high tensile strength and corrosion resistance. Fully enclosed stainless steel springs provide smooth operation. Three-prong clamps are available in single and dual adjustment. The small clamp has a 1¿ (25mm) maximum grip size with a 1/4¿ (6mm) O.D. extension arm. The medium clamp has a 1 1/2¿ (38mm) maximum grip size with a 1/2¿ (13mm) O.D. extension arm. The large clamp has a 2 1/2¿ (64mm) maximum grip size with a 5/8¿ (16mm) O.D. extension arm. Complete with removable vinyl and fiberglass covers for easy cleaning.