Learn More
OHAUS Small Ampule/Tube Attachment
Catalog No. 01804352 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-352 Shaker Attachment
1 options
Catalog No. 01-804-352 Supplier OHAUS Supplier No. 83041389
Add to Cart
Add to Cart
The Small Ampule/Tube Attachment can be used with the Vortex Genie 2 family of mixers and the Vortex Genie Pulse.
  • It provides vigorous end-to-end agitation combined with vortexing of up to four 15 mm-17 mm diameter tubes or ampules.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment