Learn More
Stoelting™ Small Animal Decapitator
SDP
Catalog No. 10000124 Shop All Stoelting Co Products
Change view
Click to view available options
Animal:
Small Animal
Catalog No. Animal
10-000-124 Small Animal
1 options
Catalog No. 10-000-124 Supplier Stoelting™ Supplier No. 51330
 Restricted Item
Add to Cart
Add to Cart
Stoelting™ Small Animal Decapitator cuts cleanly and quickly through bone and tissue and is suitable for decapitating rats, mice and other small animals.
  • Hardened and ground surgical stainless steel blades
  • Aluminum base
  • Stainless steel hardware
  • 1.5 in. square maximum blade opening
  • Base has four holes for bench mounting
Specifications
For Use With (Application) Neuroscience
Material Aluminum Base, Stainless Steel Blades
Animal Small Animal
Includes 4 holes for mounting