Learn More
Braintree Scientific Small Animal Ear Tags # 2001-3000 (1,000 pieces)
SDP
Supplier:  Braintree Scientific EP20013000
Encompass
Small Animal Ear Tag - Stainless steel applicator is designed to seal tags with minimal pressure, while providing a secure seal. Applicator can be used with or without the spring. Spring applies slight closing pressure to help hold tag in applicator prior to sealing. Custom numbering available. Call for details. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4753