Learn More
Med Vet International Small Animal Feeding Tube, J0347B, 10Fr x 16in, 5 Count
Catalog No. 5000581910
Encompass_Preferred
Red latex rubber with two opposing lateral eyes. X-ray opaque. Sterile. Improved over red vinyl tubes because it is bio-compatible and allows more patient comfort with less rigidity. Can be reused and reautoclaved. 40cm (16"). 5/pk.
Catalog No. 50-005-81910 Supplier Med Vet International Supplier No. JOR347B
Red latex rubber with two opposing lateral eyes. X-ray opaque. Sterile. Improved over red vinyl tubes because it is bio-compatible and allows more patient comfort with less rigidity. Can be reused and reautoclaved. 40cm (16"). 5/pk.