Learn More
Med Vet International Small Animal Nasogastric Feeding Tube 3.5fr x 15", J0348, 5/Pack
Catalog No. 5000581814
Encompass_Preferred
Small Animal Nasogastric Feeding Tube. These small animal feeding tubes are useful in longer term nasogastric oral supplementation. Transparent PVC. Rounded closed tip with two opposing lateral eyes. Luer syringe connector with attached plug. Sterile. 5/pk. J0348 Tube. 3.5fr x 15". Cat and dogs. New smaller size that can be subbed for the hard-to-get red rubber feeding tube in this small size.
Catalog No. 50-005-81814 Supplier Med Vet International Supplier No. JOR348
Small Animal Nasogastric Feeding Tube. These small animal feeding tubes are useful in longer term nasogastric oral supplementation. Transparent PVC. Rounded closed tip with two opposing lateral eyes. Luer syringe connector with attached plug. Sterile. 5/pk. J0348 Tube. 3.5fr x 15". Cat and dogs. New smaller size that can be subbed for the hard-to-get red rubber feeding tube in this small size.