Learn More
Med Vet International Small Animal Nasogastric Feeding Tubes, Sterile, J0348B,, 8Fr x 15", 5 Count
Catalog No. 5000581912
Encompass_Preferred
These small animal feeding tubes are useful in longer term nasogastric oral supplementation. Transparent plastic. Rounded closed tip with two opposing lateral eyes. Luer syringe connector with attached plug.
Catalog No. 50-005-81912 Supplier Med Vet International Supplier No. JOR348B
These small animal feeding tubes are useful in longer term nasogastric oral supplementation. Transparent plastic. Rounded closed tip with two opposing lateral eyes. Luer syringe connector with attached plug.