Learn More
Stoelting™ Small Bird Adaptor Kit
SDP
Catalog No. 10000150 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-150 Neuroscience
1 options
Catalog No. 10-000-150 Supplier Stoelting™ Supplier No. 51623
Add to Cart
Add to Cart
Uses a beak support and V-shaped beak clamp to hold the beak down and centered, in place of the incisor bar/nose clamp arrangement used for rats
Includes the 45° ear bars (nontraumatic) required for birds
Specifications
For Use With (Application) Neuroscience