Learn More
B BRAUN MED INC OEM SMALL BORE EXTENSION SET WITH
Catalog No. NC1501242
Encompass_Preferred
Female luer connector, non-removable slide clamp, distal SPIN-LOCK connector. DEHP-free, Latex-free. Priming volume: 0.5 mL. Length 13 in. (31.0 cm)
** Please note that one of the following licenses is required in order to process an order:
¿Board of Pharmacy
¿Medical Director
¿Drug/Wholesale Distributor
This item is not returnable. View return policy
Catalog No. NC1501242 Supplier B BRAUN MED INC OEM Supplier No. 473105
Female luer connector, non-removable slide clamp, distal SPIN-LOCK connector. DEHP-free, Latex-free. Priming volume: 0.5 mL. Length 13 in. (31.0 cm)
** Please note that one of the following licenses is required in order to process an order:
¿Board of Pharmacy
¿Medical Director
¿Drug/Wholesale Distributor