Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Small Cartridges for Multichannel Cartridge Pump Head with Reduced Pulsation for Microbore 2-stop Tubing
Catalog No. NC9804980
Encompass
Cartridges feature accurate flow rates and adjustable tubing retainers to hold tubing in place ¿ for large cartridges, set tubing retainers to your tube size and you are ready to pump. On small cartridges, optimum retention is preset ¿ simply load tubing and start pumping. Cartridges are constructed from polycarbonate and glass-filled nylon. Mix small and large cartridges on pump heads that accept both sizes; two small cartridges occupy the same space as one large cartridge.
Catalog No. NC9804980 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0751985
Cartridges feature accurate flow rates and adjustable tubing retainers to hold tubing in place ¿ for large cartridges, set tubing retainers to your tube size and you are ready to pump. On small cartridges, optimum retention is preset ¿ simply load tubing and start pumping. Cartridges are constructed from polycarbonate and glass-filled nylon. Mix small and large cartridges on pump heads that accept both sizes; two small cartridges occupy the same space as one large cartridge.