Learn More
Beckman Coulter CleanSEQ Dye-Terminator Removal Kit, 8 mL
Catalog No. NC9044757
Encompass
A rapid, high performance dye-terminator removal process based on the Agencourt SPRI technology. The paramagnetic bead format requires no centrifugation or filtration and is easily performed manually or fully automated for high throughput dye-terminator removal. Compared to similar systems, the CleanSEQ system produces sequences with longer Phred 20 read lengths and higher signal intensities than any other purification technology.
Catalog No. NC9044757 Supplier Beckman Coulter Supplier No. A29151
A rapid, high performance dye-terminator removal process based on the Agencourt SPRI technology. The paramagnetic bead format requires no centrifugation or filtration and is easily performed manually or fully automated for high throughput dye-terminator removal. Compared to similar systems, the CleanSEQ system produces sequences with longer Phred 20 read lengths and higher signal intensities than any other purification technology.