Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Memmert Memmert CO2 Incubator ICO 50, 115Volt, +5C above Ambient to +50C, Humidity 40 to 97%, CO2 0 to 20%, including K7
SDP

Catalog No. 5000451119
Encompass_Preferred

Memmert CO2 Incubator ICO 50, 115Volt, +5C above Ambient to +50C, Humidity 40 to 97%, CO2 0 to 20%, including K7

Catalog No. 50-004-51119 Supplier Memmert Supplier No. ICO 50K7 115V
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Memmert CO2 Incubator ICO 50, 115Volt, +5C above Ambient to +50C, Humidity 40 to 97%, CO2 0 to 20%, including K7

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.