Learn More
Chemglass Life Sciences Small Condenser Only, Extraction Apparatus, Soxhlet, 34/45
SDP
Catalog No. CG136804
Encompass_Preferred
Small condenser. Part of the CG1368 series. Condenser has an enlarged uptake tube to improve condensing capacity.
Catalog No. CG136804 Supplier Chemglass Life Sciences Supplier No. CG136804
Small condenser. Part of the CG1368 series. Condenser has an enlarged uptake tube to improve condensing capacity.