Learn More
Genhunter Corporation PerfectWestern, Small, Clear
Catalog No. NC9707318
Encompass
Small Container, Clear,For Mini-Protein gels cut in half,Dimensions (LxWxH): 7.3 x 5.1 x 3.3 cm,The clear container allows easy visualization of the blot
Catalog No. NC9707318 Supplier Genhunter Corporation Supplier No. B104
Small Container, Clear,For Mini-Protein gels cut in half,Dimensions (LxWxH): 7.3 x 5.1 x 3.3 cm,The clear container allows easy visualization of the blot