Learn More
Electron Microscopy Sciences Small Cryo Storage Box
SDP
Catalog No. 50996441
Encompass_Preferred
Made from heavy-duty chipboard. These boxes store vials neatly and fit most standard rack systems. They come with dividers and are slotted for cross-ventilation. The small box is 5 1/2" (132 mm) square, 2" (50 mm) high and contains divided spaces for up to 81 2 ml vials. 5.5 " square, 2" high with divided spaces for up to 81 2ml vials.
Catalog No. 50-996-441 Supplier Electron Microscopy Sciences Supplier No. 6201010
Made from heavy-duty chipboard. These boxes store vials neatly and fit most standard rack systems. They come with dividers and are slotted for cross-ventilation. The small box is 5 1/2" (132 mm) square, 2" (50 mm) high and contains divided spaces for up to 81 2 ml vials. 5.5 " square, 2" high with divided spaces for up to 81 2ml vials.