Learn More
Chemgenes Corporation Small Drying Traps, 10 pack, effective for 10-50 mL acetonitrile solutions, reduces moisture from 250 ppm to below 50 ppm, pre-activated, vacuum sealed
SDP
Catalog No. NC1738854
Encompass
The Small Drying Traps are designed to maintain highly anhydrous environments essential for high-quality oligonucleotide synthesis. These traps effectively dry acetonitrile solutions ranging from 10-50 mL, significantly reducing moisture content from approximately 250 ppm to below 50 ppm. They come pre-activated and are packaged in vacuum-sealed foil packets for immediate usability. These traps are crucial for improving coupling efficiency during DNA/RNA synthesis, making them a vital part of laboratory workflows.
  • Creates super anhydrous conditions critical for DNA/RNA synthesis
  • Reduces moisture in acetonitrile from 250 ppm to below 50 ppm
  • Supplied fully activated for immediate use
  • Compatible with various acetonitrile-based solutions
  • Enhances coupling yields during oligonucleotide synthesis
  • Packaged in vacuum-sealed foil packets
  • 10 traps per pack
Catalog No. NC1738854 Supplier Chemgenes Corporation Supplier No. DMT1975
The Small Drying Traps are designed to maintain highly anhydrous environments essential for high-quality oligonucleotide synthesis. These traps effectively dry acetonitrile solutions ranging from 10-50 mL, significantly reducing moisture content from approximately 250 ppm to below 50 ppm. They come pre-activated and are packaged in vacuum-sealed foil packets for immediate usability. These traps are crucial for improving coupling efficiency during DNA/RNA synthesis, making them a vital part of laboratory workflows.
  • Creates super anhydrous conditions critical for DNA/RNA synthesis
  • Reduces moisture in acetonitrile from 250 ppm to below 50 ppm
  • Supplied fully activated for immediate use
  • Compatible with various acetonitrile-based solutions
  • Enhances coupling yields during oligonucleotide synthesis
  • Packaged in vacuum-sealed foil packets
  • 10 traps per pack