Learn More
Saf T Pak Inc STP-700 - Disposable 2 - Part Secondary Pressure Vessel, Small, 50/Case
SDP
Catalog No. NC9198944 Shop All Saf-T-Pak Inc. Products
Encompass_Preferred
Vessel; Pressure; 2 Part Secondary; Small; Disposable; Inner leak proof polybag 4.75 x 8.5in.; Tyvek outer envelope 3.75 x 6.5in.; 10mL vials, 50/Case
Catalog No. NC9198944 Supplier Saf T Pak Inc Supplier No. STP700
Vessel; Pressure; 2 Part Secondary; Small; Disposable; Inner leak proof polybag 4.75 x 8.5in.; Tyvek outer envelope 3.75 x 6.5in.; 10mL vials, 50/Case