Learn More
Electron Microscopy Sciences Flat Embedding Mold Small Flat Embedding Mold, 4 mm deep, 10 x 6 mm
SDP
Catalog No. 50930535
Encompass_Preferred
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath. This mold produces 12 embedding blocks with tapered ends.
Depth: 4 mm
Size: 10 x 6 mm
Cavities: 12
Catalog No. 50-930-535 Supplier Electron Microscopy Sciences Supplier No. 70903
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath. This mold produces 12 embedding blocks with tapered ends.
Depth: 4 mm
Size: 10 x 6 mm
Cavities: 12