Learn More
Mitegen Small Foam Dewar, 500 mL Capacity, Lightweight, Shatterproof, Excellent Thermal Insulation for Lab Chilling
SDP
Catalog No. 501187302
Encompass
This Small Foam Dewar provides a versatile and safe solution for various laboratory applications demanding precise temperature maintenance for sensitive materials on a smaller scale. With a convenient 500 mL (0.5 Liter) capacity, this dewar is ideally suited for handling small amounts of liquid nitrogen, dry ice, ice, or other cooling agents. Constructed from robust, high-density foam, it offers excellent thermal insulation, crucial for maintaining stable temperatures and ensuring the integrity of your samples or reagents. Its lightweight and inherently shatterproof design enhances safety in the lab, minimizing risks associated with accidental drops compared to traditional glass vessels. Featuring a wide, accessible opening and a stable base, this foam dewar is a reliable and indispensable addition for quick chilling, temporary cryogenic storage, and other small-scale, temperature-sensitive processes.
Catalog No. 50-118-7302 Supplier Mitegen Supplier No. MFD500
This Small Foam Dewar provides a versatile and safe solution for various laboratory applications demanding precise temperature maintenance for sensitive materials on a smaller scale. With a convenient 500 mL (0.5 Liter) capacity, this dewar is ideally suited for handling small amounts of liquid nitrogen, dry ice, ice, or other cooling agents. Constructed from robust, high-density foam, it offers excellent thermal insulation, crucial for maintaining stable temperatures and ensuring the integrity of your samples or reagents. Its lightweight and inherently shatterproof design enhances safety in the lab, minimizing risks associated with accidental drops compared to traditional glass vessels. Featuring a wide, accessible opening and a stable base, this foam dewar is a reliable and indispensable addition for quick chilling, temporary cryogenic storage, and other small-scale, temperature-sensitive processes.