Learn More
World Precision Instrument Small FORT Force Transducers-FORT10g
SDP
Catalog No. 502521179
Encompass
The FORT Force Transducers are reliable tools for high precision force measurement. Using balanced semiconductor strain gauges, the force transducer produces linear output voltage versus applied force input with very little deflection. It is a temperature-compensated, full-bridge configuration with four high sensitivity semiconductor strain gauges. It has a broad dynamic measuring range and a very high sensitivity.
Catalog No. 50-252-1179 Supplier World Precision Instrument Supplier No. FORT10G
The FORT Force Transducers are reliable tools for high precision force measurement. Using balanced semiconductor strain gauges, the force transducer produces linear output voltage versus applied force input with very little deflection. It is a temperature-compensated, full-bridge configuration with four high sensitivity semiconductor strain gauges. It has a broad dynamic measuring range and a very high sensitivity.