Learn More
Med Vet International Small Fragment Forceps, Large, 8in, J0212L, Each
Catalog No. 5000585484
Encompass_Preferred
5 1/4". Very small fragments (e.g., malleolar fractures in cats) require appropriate instrumentation. The fine ratchet on these forceps allows very precise closure. The tips are stepped down to very accurate reduction without over penetration.
Catalog No. 50-005-85484 Supplier Med Vet International Supplier No. JOR212L
5 1/4". Very small fragments (e.g., malleolar fractures in cats) require appropriate instrumentation. The fine ratchet on these forceps allows very precise closure. The tips are stepped down to very accurate reduction without over penetration.