Learn More
Ampco Small Gasket Knife with Looped End
SDP
Catalog No. 19174138 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
27 in.
Catalog No. Length (English)
19-174-138 27 in.
1 options
Catalog No. 19-174-138 Supplier Ampco Supplier No. K52
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant gasket knife.
  • Please see image for additional information regarding measurements.
  • Tip width (a): 1 and 3/16 in.
  • Tip length (b): 15 and 3/4 in.
  • Overall length: 27 in.
Specifications
Product Type Gasket Knife
Length (English) Blade 15 -3/4 in.
Length (English) 27 in.
Quantity 1 Ea.