Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Small Group Learning: Introduction to Microbiology
GREENER_CHOICESCI_ED

Catalog No. S280003 Shop All Aldon Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S280003 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S280003 Supplier Innovating Science™ Supplier No. IS3014SGL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Small Group Learning: Introduction to Microbiology kit will allow students to collect and grow bacteria and then learn techniques for studying the bacteria they have grown.

  • Bacteria, good and bad, is all around us.
  • In this activity students will collect and grow bacteria and then learn techniques for studying the bacteria they have grown.
  • They will perform a simple staining technique on bacterial cells to study the morphology.
  • They will also perform a differential staining technique, Gram staining, on bacterial cells and determine if the collected bacteria are Gram-positive, Gram-negative, or both.
  • Kit contains enough materials for 5 groups of students.
  • Teacher's Manual and Student Study Guide copymasters are included.

Specifications

Certifications/Compliance Next Generation Science Standards (NGSS)
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Life Science
Includes 50 mL Prepared Nutrient Agar, 10 mL Methylene Blue, 10 mL Crystal Violet, 10 mL Gram’s Iodine, 10 mL Safranin O, 30 mL Ethanol, 95, Microscope Slides (72 Pk.), Coverslips, 5 Sterile Petri Dishes, 5 Sterile Cotton Swabs (2 Pk.)
Class Size 5 Lab Groups
Product Type Microbiology Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.