Learn More
Electron Microscopy Sciences Small Handle Needle File
SDP
Catalog No. 5030647
Encompass_Preferred
Blue anodized aluminum handles are well knurled with an excellent grip. The length of the handle is 4" and features a steel easy-release chuck, which simply presses into the barrel and remains secure. Handle will last a lifetime Made in the USA. Length: 4", diameter: 3/8".
Catalog No. 50-306-47 Supplier Electron Microscopy Sciences Supplier No. 6221301
Blue anodized aluminum handles are well knurled with an excellent grip. The length of the handle is 4" and features a steel easy-release chuck, which simply presses into the barrel and remains secure. Handle will last a lifetime Made in the USA. Length: 4", diameter: 3/8".