Learn More
Braintree Scientific Heated Hard Pad, 6"""" x 8""""
SDP
Supplier:  Braintree Scientific HHP05
Encompass
Water Heated Hard Pads - Connects to Gaymar, Androit and other pumps. Durable polycarbonate construction. Scalpel, needle, and towel clamp proof. Smooth surfaces are easy to clean helping to prevent nosocomial infection. Highly puncture resistant. **Sold for research purposes only and not for human use**
Catalog No. NC9979408
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.