Learn More
Med Vet International Small Jacobs Chuck, J0214
Catalog No. 5000583101
Encompass_Preferred
Lightweight blue anodized aluminum. 120g. 14cm.Much finer, more delicate, and less than half the weight of standard chucks Ideal for small dogs and cats when using "k" wires and small pins up to 5/32"Handle not slotted as JOR214L
Catalog No. 50-005-83101 Supplier Med Vet International Supplier No. JOR214
Lightweight blue anodized aluminum. 120g. 14cm.Much finer, more delicate, and less than half the weight of standard chucks Ideal for small dogs and cats when using "k" wires and small pins up to 5/32"Handle not slotted as JOR214L