Learn More
Chemglass Life Sciences Small Universal Kwik Klamp II
SDP
Supplier:  Chemglass Life Sciences CG925310
Encompass_Preferred
Small Universal Kwik Klamp II. Universal Kwik Klamp II is constructed of aluminum that has a chemically resistant Hardcote and PTFE anodized finish. Versatile design can be used on 1/2” OD Rod. The small Klamp attaches to 3/4” pipe (1.050” OD) and 1" pipe (1.315" OD). Useful in attaching temperature monitors or controllers, additional supports and clamps. The optional CG-3498-03 1/2” OD rod, bent 90° is necessary when supporting a temperature monitor or controller to a reactor stand or laboratory hood lattice.Features:• Heavy Duty Design• Corrosion Resistant• Large Clamping Knobs• Universal Design Securely Fastens To 10L Through 100L Reactor Frames And Stationary Or Mobile Benchtop Stands• 1/2" Diameter Rod Commonly Used For Lattice In Hoods• Clamps and Support Rods Can Be Mounted Horizontally Or Vertically• Secure Up To Four Different Rods, Clamps Or SupportsSmall Kwik Klamp II for use with: CG-1950, CG-1951, CG-1965-X, and CG-1968-F Support Frame
Catalog No. 50-871-980