Learn More
Electron Microscopy Sciences High Opacity Paint Markers Small Black
SDP
Catalog No. 50949074
Encompass_Preferred
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface.Weatherproof. Resistant to alcohol and acetone. Contain xylene.
Catalog No. 50-949-074 Supplier Electron Microscopy Sciences Supplier No. 72168B1
Constructed aluminum barrel, actuating ball and hard felt tips, these markers permanently cover nearly any surface.Weatherproof. Resistant to alcohol and acetone. Contain xylene.