Learn More
Bellco Glass, Inc. Small Mesh,10x15cm-5 Pkg
SDP
Supplier:  Bellco Glass, Inc. 791015005
Encompass_Preferred
Used for auto blot hybridization ovens --Pore size: 125 microns open area; 39%--Mesh count: (per inch); 127--Weave type: 1/1 plain fabric--Thickness: 120 microns--Weight: 1.5 Ounces per sq yd Size: 15x10cm. MADE IN THE USA
Catalog No. 50-194-6509