Learn More
Marlin Steel Wire Products Small Mesh Basket w/ Latch and Lid
SDP
Catalog No. NC1959433
Encompass_Preferred
Dimensions - 3.39"L x 2.75"W x 3.53"HSmall mesh basket ideal for material handling, washing, cleaning, sterilization, and greasing. Latch and lid ensures everything stays inside.Electroplish finish for smooth, shiny finish and stong nonstick properties6 mesh openings per linear inch
Catalog No. NC1959433 Supplier Marlin Steel Wire Products Supplier No. 000027800131
Dimensions - 3.39"L x 2.75"W x 3.53"HSmall mesh basket ideal for material handling, washing, cleaning, sterilization, and greasing. Latch and lid ensures everything stays inside.Electroplish finish for smooth, shiny finish and stong nonstick properties6 mesh openings per linear inch