Learn More
Marlin Steel Wire Products Mesh Basket with Locking Lid 3.20"D x 12"H
SDP
Catalog No. NC1959483
Encompass_Preferred
Stainless steel mesh baskets with a lid that locks30 mesh openings per linear inchElectropolish finish for smooth, shiny finish and nonstick properties.
Catalog No. NC1959483 Supplier Marlin Steel Wire Products Supplier No. 000036323331
Stainless steel mesh baskets with a lid that locks30 mesh openings per linear inchElectropolish finish for smooth, shiny finish and nonstick properties.