Learn More
Marlin Steel Wire Products Small Mesh Basket with Lid
SDP
Catalog No. NC1959434
Encompass_Preferred
Dimensions - 14"L x 10"W x 6"HMesh basket ideal for material handling, washing, cleaning, sterilization, and greasing. Latch and lid ensures everything stays inside.rugged construction also ensures baslet longevity in applications such as dipping or moving components in medical settings.Electroplish finish for smooth, shiny finish and stong nonstick properties4 mesh openings per linear inch
Catalog No. NC1959434 Supplier Marlin Steel Wire Products Supplier No. 000030400231
Dimensions - 14"L x 10"W x 6"HMesh basket ideal for material handling, washing, cleaning, sterilization, and greasing. Latch and lid ensures everything stays inside.rugged construction also ensures baslet longevity in applications such as dipping or moving components in medical settings.Electroplish finish for smooth, shiny finish and stong nonstick properties4 mesh openings per linear inch